(4-Nitrophenyl)acetonitril ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Nitrobenzylcyanid

A14821.18 A14821.30
A14821.18EA 38.5 EUR
A14821.18 A14821.30
(4-Nitrophenyl)acetonitril ≥98%
(4-Nitrophenyl)acetonitril

Purity: 98% CAS No.: 555-21-5 Molecular Formula: C8H6N2O2 Formula Weight: 162.15 Melting Point: 114-117° Boiling Point: 195-197°/12mm Merck Reference: 14,6590 UN No.: 3439 MDL No.: MFCD00007372

Formel: C₈H₆N₂O₂
Molecular Weight: 162.15 g/mol
Siedepunkt: 195…197 °C (12 mmHg)
Schmelzpunkt: 114…117 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007372
CAS-Nummer: 555-21-5
EINECS: 209-085-0
UN: 3439
ADR: 6.1 III
Merck Index: 12,06688

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR