5-Brom-2-furancarbonsäure ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

A14840.18 A14840.30
A14840.18EA 62.1 EUR
A14840.18 A14840.30
5-Brom-2-furancarbonsäure ≥99%
5-Bromo-2-furoic acid

Purity: 99% CAS No.: 585-70-6 Molecular Formula: C5H3BrO3 Formula Weight: 190.99 Melting Point: 188-192° Beilstein Reference: 118354

Formel: C₅H₃BrO₃
Schmelzpunkt: 188…192 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003239
CAS-Nummer: 585-70-6
EINECS: 209-559-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR