Hydrochinonmonomethylether ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Hydroxyanisol, 4-Methoxyphenol, Mequinol, MEHQ, 1-Hydroxy-4-methoxybenzol

A14845.0B A14845.18 A14845.30 AB117737.0250
A14845.0BEA 112 EUR
A14845.0B A14845.18 A14845.30 ABCRAB117737.0250
Hydrochinonmonomethylether ≥98%
Hydrochinonmonomethylether

Purity: 98+% CAS No.: 150-76-5 Molecular Formula: C7H8O2 Formula Weight: 124.14 Flash Point: 132°(269°F) Melting Point: 55-58° Boiling Point: 243-246° Beilstein Reference: 507924 MDL No.: MFCD00002332

Formel: C₇H₈O₂
Siedepunkt: 243…246 °C
Schmelzpunkt: 55…58 °C
Dichte: 1,550
Flammpunkt: 132 °C (269 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002332
CAS-Nummer: 150-76-5
EINECS: 205-769-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR