Thioanisol ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 99%
CAS No.: 100-68-5
Molecular Formula: C7H8S
Formula Weight: 124.21
Flash Point: 57°(135°F)
Refractive Index: 1.5860
Odour: Stench
Melting Point: -15°
Boiling Point: 188-189°
Beilstein Reference: 1904179
UN No.: 1993
MDL No.: MFCD00008559
Formel:
C₇H₈S Siedepunkt: 188…189 °C Schmelzpunkt: –15 °C Dichte: 1,058 Flammpunkt: 73 °C (163 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008559 CAS-Nummer: 100-68-5 EINECS: 202-878-2 UN: 3334 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...