Carbenicillin Dinatriumsalz
Lieferant: PanReac AppliChem
|
Gefahr
|
A1491.0010EA
204
EUR
A1491.0010
A1491.0005
Carbenicillin Dinatriumsalz
Carbenicillin Dinatriumsalz
Formel:
C₁₇H₁₆N₂O₆S·2Na Molecular Weight: 422,363 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00077683 CAS-Nummer: 4800-94-6 EINECS: 225-360-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...