Trimethylamin-N-oxid Dihydrat ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Trimethylamine oxide dihydrate
A14916.14
A14916.06
A14916.22
A14916.36
A14916.14EA
41.8
EUR
A14916.14
A14916.06
A14916.22
A14916.36
Trimethylamin-N-oxid Dihydrat ≥98%
Trimethylamin-N-oxid Dihydrat
Purity: 98+%
CAS No.: 62637-93-8
Molecular Formula: (CH3)3NO.2H2O
Formula Weight: 111.14 (75.11anhy)
Melting Point: 96-101°
Beilstein Reference: 3612927
Merck Reference: 14,9710
MDL No.: MFCD00149077
Formel:
C₃H₉NO·2H₂O Molecular Weight: 111.14 g/mol (75.11 anhy) Schmelzpunkt: 96…101 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149077 CAS-Nummer: 62637-93-8 EINECS: 214-675-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...