Gentamicinsulfat
Lieferant: PanReac AppliChem
|
Gefahr
|
Synonyme:
Gentamicin Sulfatsalz
A1492.0005
A1492.0025
A1492.0005EA
38.3
EUR
A1492.0005
A1492.0025
Gentamicinsulfat
Gentamicinsulfat
Schmelzpunkt:
218…237 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00270181 CAS-Nummer: 1405-41-0 EINECS: 215-778-9 Merck Index: 13,04403 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...