Bicin ≥99%
Lieferant: Thermo Fisher Scientific
Synonyme:
N,N-Bis(2-hydroxyethyl)glycin
A14957.0B
A14957.18
A14957.30
A14957.0BEA
463
EUR
A14957.0B
A14957.18
A14957.30
Bicin ≥99%
Bicin
Purity: 99%
CAS No.: 150-25-4
Molecular Formula: C6H13NO4
Formula Weight: 163.17
pH: 4-5
Melting Point: ca 191° dec.
Beilstein Reference: 1769362
Merck Reference: 14,1201
MDL No.: MFCD00004295
Good's buffers
Formel:
C₆H₁₃NO₄ Schmelzpunkt: ∼ 191 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004295 CAS-Nummer: 150-25-4 EINECS: 205-755-1 Merck Index: 14,1201 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...