4-Methoxybenzylamin ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 98+%
CAS No.: 2393-23-9
Molecular Formula: C8H11NO
Formula Weight: 137.18
Flash Point: 114°(237°F)
Refractive Index: 1.5460
Odour: Amine-like
Melting Point: -10°
Boiling Point: 236-237°
Beilstein Reference: 508206
UN No.: 2735
MDL No.: MFCD00008122
Formel:
C₈H₁₁NO Siedepunkt: 236…237 °C Schmelzpunkt: –10 °C Dichte: 1,053 Flammpunkt: 114 °C (237 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008122 CAS-Nummer: 2393-23-9 EINECS: 219-247-2 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...