7-Amino-4-methylcumarin 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
7-Amino-4-methyl-2-benzopyron, AMC
A15017.03
A15017.06
A15017.MD
A15017.03EA
111
EUR
A15017.03
A15017.06
A15017.MD
7-Amino-4-methylcumarin 98%
7-Amino-4-methylcumarin
Purity: 98%
CAS No.: 26093-31-2
Molecular Formula: C10H9NO2
Formula Weight: 175.19
Melting Point: 225-228°
Beilstein Reference: 142231
Formel:
C₁₀H₉NO₂ Siedepunkt: 352 °C (1013 hPa) Schmelzpunkt: 225…228 °C Dichte: 1,377 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006868 CAS-Nummer: 26093-31-2 EINECS: 247-454-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...