4-Amino-2,6-dichlorpyridin ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2,6-Dichlor-4-pyridylamin, 2,6-Dichlorpyridin-4-amin

A15030.06
A15030.06EA 121 EUR
A15030.06
4-Amino-2,6-dichlorpyridin ≥98%
4-Amino-2,6-dichlorpyridin

Purity: 98+% CAS No.: 2587-02-2 Molecular Formula: C5H4Cl2N2 Formula Weight: 163.01 Odour: Unpleasant Melting Point: 172-176° Beilstein Reference: 114351 MDL No.: MFCD00052832

Formel: C₅H₄Cl₂N₂
Molecular Weight: 163,01 g/mol
Schmelzpunkt: 172…176 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052832
CAS-Nummer: 2587-02-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR