N-Ethylanilin ≥97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: N-Ethyl-N-phenylamine

A15060.0F
A15060.0FEA 117 EUR
A15060.0F
N-Ethylanilin ≥97%
N-Ethylanilin

Purity: 97% CAS No.: 103-69-5 Molecular Formula: C8H11N Formula Weight: 121.18 Flash Point: 85°(185°F) Refractive Index: 1.5540 Odour: Odourless Melting Point: -64° Boiling Point: 204-205° Beilstein Reference: 507468 Merck Reference: 14,3764 UN No.: 2272 MDL No.: MFCD00009025

Formel: C₈H₁₁N
Molecular Weight: 121,18 g/mol
Siedepunkt: 204…205 °C
Schmelzpunkt: –64 °C
Dichte: 0,961
Flammpunkt: 85 °C (185 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009025
CAS-Nummer: 103-69-5
EINECS: 203-135-5
UN: 2272
ADR: 6.1,III
Merck Index: 14,3764

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR