2-Brombenzaldehyd ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A15065.14
A15065.22
A15065.36
A15065.14EA
65.9
EUR
A15065.14
A15065.22
A15065.36
2-Brombenzaldehyd ≥98%
2-Brombenzaldehyd
Purity: 98%
CAS No.: 6630-33-7
Molecular Formula: C7H5BrO
Formula Weight: 185.03
Flash Point: 95°(203°F)
Refractive Index: 1.5960
Odour: Characteristic
Melting Point: 18-21°
Boiling Point: 228-230°
Beilstein Reference: 636132
MDL No.: MFCD00003300
Formel:
C₇H₅BrO Siedepunkt: 228…230 °C Schmelzpunkt: 18…21 °C Dichte: 1,597 Flammpunkt: 95 °C (203 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003300 CAS-Nummer: 6630-33-7 EINECS: 229-622-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...