(1-Hexadecyl)triphenylphosphoniumbromid ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A15180.06
A15180.09
A15180.22
A15180.06EA
25.6
EUR
A15180.06
A15180.09
A15180.22
(1-Hexadecyl)triphenylphosphoniumbromid ≥98%
(1-Hexadecyl)triphenylphosphoniumbromid
Purity: 98+%
CAS No.: 14866-43-4
Molecular Formula: CH3(CH2)15P(C6H5)3Br
Formula Weight: 567.64
Melting Point: 99-101°
Beilstein Reference: 3582592
Formel:
H₃C(CH₂)₁₅P(C₆H₅)₃Br Molecular Weight: 567.63 g/mol Schmelzpunkt: 99…101 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051858 CAS-Nummer: 14866-43-4 EINECS: 238-939-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...