Pamoic acid 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 99%
CAS No.: 130-85-8
Molecular Formula: C23H16O6
Formula Weight: 388.38
Melting Point:
Applications: Used in preparation of repository derivatives of medicinal agents
Formel:
C₂₃H₁₆O₆ Schmelzpunkt: >300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004079 CAS-Nummer: 130-85-8 EINECS: 204-998-0 Merck Index: 14,7005 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...