2-Phenylethanol ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 98+%
CAS No.: 60-12-8
Molecular Formula: C8H10O
Formula Weight: 122.17
Flash Point: 101°(214°F)
Physical Form: Liquid
Density: 1.021
Refractive Index: 1.5330
Appearance: Colourless
Melting Point: -27°
Boiling Point: 219°
Beilstein Reference: 1905732
Merck Reference: 14,7224
UN No.: 2810
MDL No.: MFCD00002886
Formel:
C₆H₅CH₂CH₂OH Siedepunkt: 219 °C Schmelzpunkt: –27 °C Dichte: 1,021 Flammpunkt: 101 °C (214 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002886 CAS-Nummer: 60-12-8 EINECS: 200-456-2 UN: 2810 ADR: 6.1,III Merck Index: 14,7224 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...