(4-Hydroxyphenyl)acetamid ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Hydroxyphenylacetamide

A15266.09 A15266.18
A15266.09EA 28.2 EUR
A15266.09 A15266.18
(4-Hydroxyphenyl)acetamid ≥99%
(4-Hydroxyphenyl)acetamid

Purity: 99% CAS No.: 17194-82-0 Molecular Formula: C8H9NO2 Formula Weight: 151.17 Melting Point: 173-175° Beilstein Reference: 2085984

Formel: C₈H₉NO₂
Schmelzpunkt: 173…175 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017145
CAS-Nummer: 17194-82-0
EINECS: 241-235-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR