(4-Hydroxyphenyl)acetamid ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-Hydroxyphenylacetamide
A15266.09
A15266.18
A15266.09EA
28.2
EUR
A15266.09
A15266.18
(4-Hydroxyphenyl)acetamid ≥99%
(4-Hydroxyphenyl)acetamid
Purity: 99%
CAS No.: 17194-82-0
Molecular Formula: C8H9NO2
Formula Weight: 151.17
Melting Point: 173-175°
Beilstein Reference: 2085984
Formel:
C₈H₉NO₂ Schmelzpunkt: 173…175 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017145 CAS-Nummer: 17194-82-0 EINECS: 241-235-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...