Bortrifluorid-Diethylether-Komplex ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

A15275.AE A15275.AP A15275.AU
A15275.AEEA 32 EUR
A15275.AE A15275.AP A15275.AU
Bortrifluorid-Diethylether-Komplex ≥98%
Bortrifluorid-Diethylether-Komplex

Purity: 98+% CAS No.: 109-63-7 Molecular Formula: C4H10BF3O Formula Weight: 141.93 Flash Point: 47°(116°F) Refractive Index: 1.3440 Odour: Acrid Melting Point: -60° Boiling Point: 125-126° Beilstein Reference: 3909607 Merck Reference: 14,1350 UN No.: 2604 MDL No.: MFCD00013194

Formel: C₄H₁₀BF₃O
Siedepunkt: 125…126 °C
Schmelzpunkt: –60 °C
Dichte: 1,130
Flammpunkt: 47 °C (116 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00013194
CAS-Nummer: 109-63-7
EINECS: 203-689-8
UN: 2604
ADR: 8,I
Merck Index: 14,1350

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR