2-Amino-5-brom-3-methylpyridin 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Amino-5-brom-3-picolin, 5-Brom-3-methyl-2-pyridylamin, 5-Brom-3-methylpyridin-2-amin

A15318.06
A15318.06EA 49.2 EUR
A15318.06
2-Amino-5-brom-3-methylpyridin 97%
2-Amino-5-brom-3-methylpyridin

Purity: 97% CAS No.: 3430-21-5 Molecular Formula: C6H7BrN2 Formula Weight: 187.04 Odour: Unpleasant Melting Point: 90-94° Beilstein Reference: 386426 MDL No.: MFCD00068232

Formel: C₆H₇BrN₂
Molecular Weight: 187,04 g/mol
Schmelzpunkt: 90…94 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00068232
CAS-Nummer: 3430-21-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR