1,4-Diiodbenzol ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A15335.14
A15335.22
A15335.14EA
53.1
EUR
A15335.14
A15335.22
1,4-Diiodbenzol ≥98%
1,4-Diiodbenzol
Purity: 98%
CAS No.: 624-38-4
Molecular Formula: C6H4I2
Formula Weight: 329.91
Melting Point: 128-132°
Boiling Point: 285°
MDL No.: MFCD00001054
Formel:
C₆H₄I₂ Molecular Weight: 329.91 g/mol Siedepunkt: 285 °C Schmelzpunkt: 128…132 °C Dichte: 2.88 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001054 CAS-Nummer: 624-38-4 EINECS: 210-842-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...