5-Fluorindol ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

A15346.03 A15346.06 A15346.14
A15346.03EA 55.2 EUR
A15346.03 A15346.06 A15346.14
5-Fluorindol ≥99%
5-Fluorindol

Purity: 99% CAS No.: 399-52-0 Molecular Formula: C8H6FN Formula Weight: 135.14 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Melting Point: 42-47° Beilstein Reference: 112350 MDL No.: MFCD00005671

Formel: C₈H₆FN
Siedepunkt: 85 °C (0,5 mmHg)
Schmelzpunkt: 42…47 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Kühlschrank
MDL: MFCD00005671
CAS-Nummer: 399-52-0
EINECS: 206-917-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR