4-Fluor-3-nitroanilin ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A15371.14
A15371.22
A15371.36
A15371.14EA
36.4
EUR
A15371.14
A15371.22
A15371.36
4-Fluor-3-nitroanilin ≥98%
4-Fluor-3-nitroanilin
Purity: 98%
CAS No.: 364-76-1
Molecular Formula: C6H5FN2O2
Formula Weight: 156.12
Flash Point: 91°(195°F)
Melting Point: 94-98°
Beilstein Reference: 2210199
MDL No.: MFCD00007833
Formel:
C₆H₅FN₂O₂ Molecular Weight: 156.12 g/mol Schmelzpunkt: 94…98 °C Dichte: 0.43 g/cm³ (20 °C) Flammpunkt: 91 °C (195 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007833 CAS-Nummer: 364-76-1 EINECS: 206-665-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...