ω-Brom-4-nitroacetophenon 95%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
2-Brom-4'-nitroacetophenon, 4-Nitrophenacylbromid, Brommethyl-p-nitrophenylketon, α-Brom-4-nitroacetophenon
A15374.09
A15374.18
A15374.09EA
37
EUR
A15374.09
A15374.18
ω-Brom-4-nitroacetophenon 95%
ω-Brom-4-nitroacetophenon
Purity: 95%
CAS No.: 99-81-0
Molecular Formula: C8H6BrNO3
Formula Weight: 244.05
Melting Point: 95-98°
Beilstein Reference: 393567
UN No.: 1759
MDL No.: MFCD00007356
Formel:
C₈H₆BrNO₃ Schmelzpunkt: 95…98 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007356 CAS-Nummer: 99-81-0 EINECS: 202-789-9 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...