ω-Brom-4-nitroacetophenon 95%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 2-Brom-4'-nitroacetophenon, 4-Nitrophenacylbromid, Brommethyl-p-nitrophenylketon, α-Brom-4-nitroacetophenon

A15374.09 A15374.18
A15374.09EA 37 EUR
A15374.09 A15374.18
ω-Brom-4-nitroacetophenon 95%
ω-Brom-4-nitroacetophenon

Purity: 95% CAS No.: 99-81-0 Molecular Formula: C8H6BrNO3 Formula Weight: 244.05 Melting Point: 95-98° Beilstein Reference: 393567 UN No.: 1759 MDL No.: MFCD00007356

Formel: C₈H₆BrNO₃
Schmelzpunkt: 95…98 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007356
CAS-Nummer: 99-81-0
EINECS: 202-789-9
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR