1,4-Diethoxybenzol ≥98%

Lieferant: Thermo Fisher Scientific

A15385.14
A15385.14EA 39.1 EUR
A15385.14
1,4-Diethoxybenzol ≥98%
1,4-Diethoxybenzene

Purity: 98% CAS No.: 122-95-2 Molecular Formula: C10H14O2 Formula Weight: 166.22 Melting Point: 69-71° Boiling Point: 245-248° MDL No.: MFCD00015146

Formel: C₁₀H₁₄O₂
Siedepunkt: 245…248 °C
Schmelzpunkt: 69…71 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00015146
CAS-Nummer: 122-95-2
EINECS: 204-585-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR