Furil 98%
Lieferant: Thermo Fisher Scientific
A15386.14
A15386.14EA
318
EUR
A15386.14
Furil 98%
Furil
Purity: 98%
CAS No.: 492-94-4
Molecular Formula: C10H6O4
Formula Weight: 190.15
Melting Point: 162-165°
Beilstein Reference: 383882
MDL No.: MFCD00003241
Formel:
C₁₀H₆O₄ Schmelzpunkt: 162…165 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003241 CAS-Nummer: 492-94-4 EINECS: 207-766-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...