Hydroxylammoniumchlorid 99%

Lieferant: Thermo Fisher Scientific
Achtung

A15398.0B A15398.0I A15398.22 A15398.36
A15398.0BEA 132 EUR
A15398.0B A15398.0I A15398.22 A15398.36
Hydroxylammoniumchlorid 99%
Hydroxylammoniumchlorid

Purity: 99% CAS No.: 5470-11-1 Molecular Formula: H3NO.HCl Formula Weight: 69.49 Odour: Acidic pH: 3.6 Melting Point: 152° dec. Beilstein Reference: 3539763 Merck Reference: 14,4828 UN No.: 2923 MDL No.: MFCD00051089

Formel: NH₂OH·HCl
Siedepunkt: 306 °C (1013 hPa)
Dichte: 1,67
Lagertemperatur: Raumtemperatur
MDL: MFCD00051089
CAS-Nummer: 5470-11-1
EINECS: 226-798-2
UN: 2923
ADR: 8,III
Merck Index: 14,4828

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR