N-Acetyl-L-cystein ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: Ac-Cys-OH, (R)-(+)-N-Acetylcystein, (R)-(+)-2-(Ethanoylamino)-3-mercaptopropansäure

A15409.14 A15409.22 A15409.36
A15409.14EA 38.2 EUR
A15409.14 A15409.22 A15409.36
N-Acetyl-L-cystein ≥98%
N-Acetyl-L-cystein

Purity: 98+% CAS No.: 616-91-1 Molecular Formula: C5H9NO3S Formula Weight: 163.20 Melting Point: 108-111° Beilstein Reference: 1724426 Merck Reference: 14,88 Optical Rotation: +5.3° (c=2 in water) MDL No.: MFCD00004880


A mucolytic agent for isolation of mycobacteria from sputum

Formel: C₅H₉NO₃S
Siedepunkt: 371 °C (1013 hPa)
Schmelzpunkt: 108…111 °C
Dichte: 1,48 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004880
CAS-Nummer: 616-91-1
EINECS: 210-498-3
Merck Index: 14,88

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR