Benzotriazol ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 99%
CAS No.: 95-14-7
Molecular Formula: C6H5N3
Formula Weight: 119.13
Flash Point: 212°(413°F)
Physical Form: Powder
Density: 1.360
Appearance: White to pale cream
Odour: Odourless
pH: 6.0-7.0
Melting Point: 96-99°
Boiling Point: 204°/15mm
Beilstein Reference: 112133
Merck Reference: 14,1108
MDL No.: MFCD00005699
Formel:
C₆H₅N₃ Siedepunkt: 204 °C (15 mmHg) Schmelzpunkt: 96…99 °C Dichte: 1,360 Flammpunkt: 212 °C (413 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005699 CAS-Nummer: 95-14-7 EINECS: 202-394-1 UN: 2811 ADR: 6.1,III Merck Index: 14,1108 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...