Benzotriazol ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1H-1,2,3-Benzotriazol, 1H-Benzotriazol, 1,2,3-Benzotriazol

A15423.18 A15423.0B A15423.30
A15423.18EA 31.9 EUR
A15423.18 A15423.0B A15423.30
Benzotriazol ≥99%
Benzotriazol

Purity: 99% CAS No.: 95-14-7 Molecular Formula: C6H5N3 Formula Weight: 119.13 Flash Point: 212°(413°F) Physical Form: Powder Density: 1.360 Appearance: White to pale cream Odour: Odourless pH: 6.0-7.0 Melting Point: 96-99° Boiling Point: 204°/15mm Beilstein Reference: 112133 Merck Reference: 14,1108 MDL No.: MFCD00005699

Formel: C₆H₅N₃
Molecular Weight: 119.13 g/mol
Siedepunkt: 204 °C (15 mmHg)
Schmelzpunkt: 96…99 °C
Dichte: 1.360
Flammpunkt: 212 °C (413 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005699
CAS-Nummer: 95-14-7
EINECS: 202-394-1
UN: 2811
ADR: 6.1 III
Merck Index: 13,01109
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0ba54cc42ef5ad9dbae6d7f118a3af1ae642287&vid=ce57b3f0d2583e32220d1e191d0202ad452e6475&prid=web&clid=SAPDAM

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR