N,N,N',N',N''-Pentamethyldiethylentriamin 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: N1-(2-(Dimethylamino)ethyl)-N1,N2,N2-trimethylethane-1,2-diamine, 1,1,4,7,7-Pentamethyldiethylenetriamine

A15433.14 A15433.22 A15433.36
A15433.14EA 48.3 EUR
A15433.14 A15433.22 A15433.36
N,N,N',N',N''-Pentamethyldiethylentriamin 98%
N,N,N',N',N''-Pentamethyldiethylentriamin

Purity: 98% CAS No.: 3030-47-5 Molecular Formula: C9H23N3 Formula Weight: 173.30 Flash Point: 72°(161°F) Refractive Index: 1.4420 Melting Point: -20° Boiling Point: 199° Beilstein Reference: 1741396 UN No.: 2922 MDL No.: MFCD00014876

Formel: C₉H₂₃N₃
Molecular Weight: 173.30 g/mol
Siedepunkt: 199 °C
Schmelzpunkt: < –20 °C
Dichte: 0.830
Flammpunkt: 72 °C (161 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00014876
CAS-Nummer: 3030-47-5
EINECS: 221-201-1
ADR: 8 II

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR