4-Cyanbenzoesäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A15434.22 A15434.06 A15434.14
A15434.22EA 360 EUR
A15434.22 A15434.06 A15434.14
4-Cyanbenzoesäure ≥98%
4-Cyanbenzoesäure

Purity: 98% CAS No.: 619-65-8 Molecular Formula: C8H5NO2 Formula Weight: 147.13 Melting Point: ca 221° dec. Beilstein Reference: 1862865

Formel: C₈H₅NO₂
Molecular Weight: 147.13 g/mol
Siedepunkt: 313
Schmelzpunkt: ∼221 °C (Zers.)
Dichte: 1.382 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002528
CAS-Nummer: 619-65-8
EINECS: 210-606-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR