7-Methoxycumarin ≥98%
Lieferant: Thermo Fisher Scientific
Achtung
|
A15441.06
A15441.14
A15441.22
A15441.06EA
39.3
EUR
A15441.06
A15441.14
A15441.22
7-Methoxycumarin ≥98%
7-Methoxycumarin
Purity: 98+%
CAS No.: 531-59-9
Molecular Formula: C10H8O3
Formula Weight: 176.17
Melting Point: 117-120°
Beilstein Reference: 141728
Formel:
C₁₀H₈O₃ Siedepunkt: 320 °C (1013 hPa) Schmelzpunkt: 117…120 °C Dichte: 1,36 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006876 CAS-Nummer: 531-59-9 EINECS: 208-513-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...