Cyanursäure ≥99%

Lieferant: Thermo Fisher Scientific

A15447.0B A15447.22 A15447.30
A15447.0BEA 111 EUR
A15447.0B A15447.22 A15447.30
Cyanursäure ≥99%
Cyanursäure

Purity: 99% CAS No.: 108-80-5 Molecular Formula: C3H3N3O3 Formula Weight: 129.08 Melting Point: 360° Beilstein Reference: 126982 Merck Reference: 14,2698 MDL No.: MFCD00082990

Formel: C₃H₃N₃O₃
Schmelzpunkt: >360 °C
Dichte: 2,5
Lagertemperatur: Raumtemperatur
MDL: MFCD00082990
CAS-Nummer: 108-80-5
EINECS: 203-618-0
Merck Index: 14,2698

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR