3,5-Bis(trifluormethyl)benzonitril ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A15460.06
A15460.14
A15460.06EA
63.2
EUR
A15460.06
A15460.14
3,5-Bis(trifluormethyl)benzonitril ≥98%
3,5-Bis(trifluormethyl)benzonitril
Purity: 98%
CAS No.: 27126-93-8
Molecular Formula: C9H3F6N
Formula Weight: 239.12
Flash Point: 72°(161°F)
Refractive Index: 1.4175
Melting Point: 16-20°
Boiling Point: 154-156°
Beilstein Reference: 3552650
UN No.: 3276
MDL No.: MFCD00000379
Formel:
C₉H₃F₆N Molecular Weight: 239.12 g/mol Siedepunkt: 154…156 °C Schmelzpunkt: 16…20 °C Dichte: 1.421 Flammpunkt: 72 °C (161 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000379 CAS-Nummer: 27126-93-8 EINECS: 248-240-7 UN: 3276 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...