2-Chlor-4-fluoranilin ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

A15492.06 A15492.14 A15492.22
A15492.06EA 40.8 EUR
A15492.06 A15492.14 A15492.22
2-Chlor-4-fluoranilin ≥97%
2-Chlor-4-fluoranilin

Purity: 97% CAS No.: 2106-02-7 Molecular Formula: C6H5ClFN Formula Weight: 145.56 Flash Point: 98°(208°F) Refractive Index: 1.5540 Odour: Characteristic Boiling Point: 191-192° Beilstein Reference: 2802559 UN No.: 2810 MDL No.: MFCD00042530

Formel: C₆H₅ClFN
Siedepunkt: 191…192 °C
Dichte: 1,219
Flammpunkt: 98 °C (208 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00042530
CAS-Nummer: 2106-02-7
EINECS: 218-282-0
UN: 2810
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR