2-Chlor-4-fluoranilin ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A15492.06
A15492.14
A15492.22
A15492.06EA
40.8
EUR
A15492.06
A15492.14
A15492.22
2-Chlor-4-fluoranilin ≥97%
2-Chlor-4-fluoranilin
Purity: 97%
CAS No.: 2106-02-7
Molecular Formula: C6H5ClFN
Formula Weight: 145.56
Flash Point: 98°(208°F)
Refractive Index: 1.5540
Odour: Characteristic
Boiling Point: 191-192°
Beilstein Reference: 2802559
UN No.: 2810
MDL No.: MFCD00042530
Formel:
C₆H₅ClFN Siedepunkt: 191…192 °C Dichte: 1,219 Flammpunkt: 98 °C (208 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042530 CAS-Nummer: 2106-02-7 EINECS: 218-282-0 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...