4-Chlor-1,3-difluorbenzol ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1-Chloro-2,4-difluorobenzene
A15495.09
A15495.18
A15495.30
A15495.09EA
30.5
EUR
A15495.09
A15495.18
A15495.30
4-Chlor-1,3-difluorbenzol ≥98%
4-Chlor-1,3-difluorbenzol
Purity: 98%
CAS No.: 1435-44-5
Molecular Formula: C6H3ClF2
Formula Weight: 148.54
Flash Point: 32°(89°F)
Refractive Index: 1.4750
Odour: Aromatic
Melting Point: -26°
Boiling Point: 126-127°
Beilstein Reference: 2081077
UN No.: 1993
MDL No.: MFCD00042569
Formel:
C₆H₃ClF₂ Molecular Weight: 148.54 g/mol Siedepunkt: 126…127 °C Schmelzpunkt: –26 °C Dichte: 1.353 Flammpunkt: 32 °C (89 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042569 CAS-Nummer: 1435-44-5 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...