4-Chlor-1,3-difluorbenzol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1-Chloro-2,4-difluorobenzene

A15495.09 A15495.18 A15495.30
A15495.09EA 30.5 EUR
A15495.09 A15495.18 A15495.30
4-Chlor-1,3-difluorbenzol ≥98%
4-Chlor-1,3-difluorbenzol

Purity: 98% CAS No.: 1435-44-5 Molecular Formula: C6H3ClF2 Formula Weight: 148.54 Flash Point: 32°(89°F) Refractive Index: 1.4750 Odour: Aromatic Melting Point: -26° Boiling Point: 126-127° Beilstein Reference: 2081077 UN No.: 1993 MDL No.: MFCD00042569

Formel: C₆H₃ClF₂
Molecular Weight: 148.54 g/mol
Siedepunkt: 126…127 °C
Schmelzpunkt: –26 °C
Dichte: 1.353
Flammpunkt: 32 °C (89 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00042569
CAS-Nummer: 1435-44-5
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR