3,5-Difluoranisol ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 1,3-Difluoro-5-methoxybenzene

A15497.14
A15497.14EA 153 EUR
A15497.14
3,5-Difluoranisol ≥97%
3,5-Difluoranisol

Purity: 97% CAS No.: 93343-10-3 Molecular Formula: C7H6F2O Formula Weight: 144.12 Flash Point: 43°(109°F) Refractive Index: 1.4660 Boiling Point: 145-148° Beilstein Reference: 4966844 UN No.: 1993 MDL No.: MFCD00042560

Formel: C₇H₆F₂O
Molecular Weight: 144,12 g/mol
Siedepunkt: 145…148 °C
Dichte: 1,234
Flammpunkt: 43 °C (109 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00042560
CAS-Nummer: 93343-10-3
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR