4'-(Trifluormethyl)acetophenon ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
p-(Trifluormethyl)acetophenon, 4-Acetylbenzotrifluorid, 1-[4-(Trifluormethyl)phenyl]ethan-1-on
A15529.06
A15529.14
A15529.06EA
54.1
EUR
A15529.06
A15529.14
4'-(Trifluormethyl)acetophenon ≥98%
4'-(Trifluormethyl)acetophenon
Purity: 98+%
CAS No.: 709-63-7
Molecular Formula: C9H7F3O
Formula Weight: 188.15
Flash Point: 84°(183°F)
Refractive Index: 1.4620
Odour: Characteristic
Melting Point: 29-35°
Boiling Point: 83-85°/9mm
Beilstein Reference: 1870425
MDL No.: MFCD00000401
Formel:
C₉H₇F₃O Siedepunkt: 83…85 °C (9 mmHg) Schmelzpunkt: 29…35 °C Dichte: 0,924 Flammpunkt: 84 °C (183 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000401 CAS-Nummer: 709-63-7 EINECS: 211-913-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...