5-Brompentylacetat ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: 5-Bromamylacetat, Essigsäure(5-bromamyl)ester, Essigsäure(5-brompentyl)ester

A15549.14 A15549.06 A15549.22
A15549.14EA 43.8 EUR
A15549.14 A15549.06 A15549.22
5-Brompentylacetat ≥98%
5-Brompentylacetat

Purity: 98% CAS No.: 15848-22-3 Molecular Formula: CH3CO2(CH2)5Br Formula Weight: 209.09 Flash Point: 110°(230°F) Refractive Index: 1.4620 Boiling Point: 109-110°/15mm Beilstein Reference: 1752109 MDL No.: MFCD00039204

Formel: C₇H₁₃BrO₂
Siedepunkt: 109…110 °C (15 mmHg)
Dichte: 1,255
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00039204
CAS-Nummer: 15848-22-3
EINECS: 239-961-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR