5-Brompentylacetat ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
5-Bromamylacetat, Essigsäure(5-bromamyl)ester, Essigsäure(5-brompentyl)ester
A15549.14
A15549.06
A15549.22
A15549.14EA
43.8
EUR
A15549.14
A15549.06
A15549.22
5-Brompentylacetat ≥98%
5-Brompentylacetat
Purity: 98%
CAS No.: 15848-22-3
Molecular Formula: CH3CO2(CH2)5Br
Formula Weight: 209.09
Flash Point:
Formel:
C₇H₁₃BrO₂ Siedepunkt: 109…110 °C (15 mmHg) Dichte: 1,255 Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00039204 CAS-Nummer: 15848-22-3 EINECS: 239-961-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...