Lanthan(III)chlorid Heptahydrat ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

A15575.0E A15575.22 A15575.36
A15575.0EEA 867 EUR
A15575.0E A15575.22 A15575.36
Lanthan(III)chlorid Heptahydrat ≥99%

Purity: 99% CAS No.: 10025-84-0 Molecular Formula: LaCl3.7H2O Formula Weight: 371.37 (245.27anhy) Odour: Odourless Melting Point: 91° dec. MDL No.: MFCD00149756

Formel: LaCl₃·7H₂O
Siedepunkt: ∼860 °C (1013 hPa)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149756
CAS-Nummer: 10025-84-0
EINECS: 233-237-5
Merck Index: 14,5363

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR