ω-Bromacetophenon ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: α-Bromacetophenon, Phenacylbromid, 2-Bromacetophenon

A15576.22 A15576.14 A15576.36
A15576.22EA 70.6 EUR
A15576.22 A15576.14 A15576.36
ω-Bromacetophenon ≥98%
ω-Bromacetophenon

Purity: 98% CAS No.: 70-11-1 Molecular Formula: C8H7BrO Formula Weight: 199.05 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Odour: Pungent Melting Point: 48-51° Boiling Point: 140°/11mm Beilstein Reference: 606474 Merck Reference: 14,1402 UN No.: 2645 MDL No.: MFCD00000195

Formel: (C₆H₅)C(O)CH₂Br
Siedepunkt: 140 °C (11 mmHg)
Schmelzpunkt: 48…51 °C
Dichte: 1,650
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Kühlschrank
MDL: MFCD00000195
CAS-Nummer: 70-11-1
EINECS: 200-724-9
UN: 2645
ADR: 6.1,II
Merck Index: 14,1402

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR