D(+)-Saccharose 99
Lieferant: Thermo Fisher Scientific
Synonyme:
Haushaltszucker, D(+)-Sucrose, Rübenzucker, Rohrzucker, 2-β-D-Fructofuranosyl-1-α-D-glucopyranosid, Sucrose, Kristallzucker
A15583.0C
A15583.0E
A15583.36
A15583.0CEA
204
EUR
A15583.0C
A15583.0E
A15583.36
D(+)-Saccharose 99
D(+)-Saccharose
Purity: 99%
CAS No.: 57-50-1
Molecular Formula: C12H22O11
Formula Weight: 342.30
Odour: Odourless
Melting Point: 186-190°
Beilstein Reference: 90825
Merck Reference: 14,8881
Optical Rotation: +66.5° (c=10 in water)
MDL No.: MFCD00006626
Formel:
C₁₂H₂₂O₁₁ Molecular Weight: 342.3 g/mol Schmelzpunkt: 186…190 °C Dichte: 1.580 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006626 CAS-Nummer: 57-50-1 EINECS: 200-334-9 Merck Index: 13,08966 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...