D(+)-Saccharose 99

Lieferant: Thermo Fisher Scientific

Synonyme: Haushaltszucker, D(+)-Sucrose, Rübenzucker, Rohrzucker, 2-β-D-Fructofuranosyl-1-α-D-glucopyranosid, Sucrose, Kristallzucker

A15583.0C A15583.0E A15583.36
A15583.0CEA 204 EUR
A15583.0C A15583.0E A15583.36
D(+)-Saccharose 99
D(+)-Saccharose

Purity: 99% CAS No.: 57-50-1 Molecular Formula: C12H22O11 Formula Weight: 342.30 Odour: Odourless Melting Point: 186-190° Beilstein Reference: 90825 Merck Reference: 14,8881 Optical Rotation: +66.5° (c=10 in water) MDL No.: MFCD00006626

Formel: C₁₂H₂₂O₁₁
Molecular Weight: 342.3 g/mol
Schmelzpunkt: 186…190 °C
Dichte: 1.580
Lagertemperatur: Raumtemperatur
MDL: MFCD00006626
CAS-Nummer: 57-50-1
EINECS: 200-334-9
Merck Index: 13,08966
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=971b1a9d1f02c06efd63ac665dfb191799fbd5ac&vid=2ddc1affd401bacf02244ae0cb620ff18e440965&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR