N-(3-Brompropyl)phthalimid ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
N-(3-Brompropyl)phthalsäureimid, N-(3-Brompropyl)-1,2-benzoldicarbonsäureimid
A15587.14
A15587.22
A15587.36
A15587.14EA
33.2
EUR
A15587.14
A15587.22
A15587.36
N-(3-Brompropyl)phthalimid ≥98%
N-(3-Brompropyl)phthalimid
Purity: 98%
CAS No.: 5460-29-7
Molecular Formula: C11H10BrNO2
Formula Weight: 268.12
Odour: Odourless
Melting Point: 72-75°
Beilstein Reference: 157547
MDL No.: MFCD00005904
Formel:
C₁₁H₁₀BrNO₂ Siedepunkt: 433 °C (1013 hPa) Schmelzpunkt: 72…75 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005904 CAS-Nummer: 5460-29-7 EINECS: 226-738-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...