1-Bromoctadecan ≥97%

Lieferant: Thermo Fisher Scientific

Synonyme: n-Octadecylbromid

A15588.0E A15588.22 A15588.36
A15588.0EEA 233 EUR
A15588.0E A15588.22 A15588.36
1-Bromoctadecan ≥97%
1-Bromoctadecan

Purity: 97% CAS No.: 112-89-0 Molecular Formula: C18H37Br Formula Weight: 333.39 Flash Point: 110°(230°F) Odour: Odourless Boiling Point: 150-152°/0.1mm Beilstein Reference: 774145 MDL No.: MFCD00000231

Formel: BrH₂C(CH₂)₁₆CH₃
Molecular Weight: 333.4 g/mol
MDL: MFCD00000231
CAS-Nummer: 112-89-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR